Invitrogen
Manufacturer Code:PA513976
Catalog # PIPA513976
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 259-288 amino acids from the C-terminal region of human PIM3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KID-1/kinase induced by depolarization; pim-3 oncogene; serine/threonine kinase Pim-3; Serine/threonine-protein kinase pim-3; Serine/threonine-protein kinase Pim-3 PIM3
Gene Aliases: BC026639; Kid1; pim-3; PIM3
UniProt ID: (Human) Q86V86, (Mouse) P58750
Entrez Gene ID: (Human) 415116, (Mouse) 223775
Molecular Function: kinase protein kinase protein kinase receptor receptor serine/threonine protein kinase receptor transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.