Invitrogen
Manufacturer Code:PA5113070
Catalog # PIPA5113070
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Serine/threonine-protein kinase pim-3 protein (278-326AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BC026639 Kid1 KID-1/kinase induced by depolarization Pim3 pim-3 pim-3 oncogene Pim-3 proto-oncogene serine/threonine kinase proviral integration site 3 serine/threonine kinase Pim-3 Serine/threonine-protein kinase pim-3; KID-1/kinase induced by depolarization; pim-3 oncogene; serine/threonine kinase Pim-3; Serine/threonine-protein kinase pim-3
Gene Aliases: BC026639; Kid1; pim-3; PIM3
UniProt ID: (Human) Q86V86, (Mouse) P58750
Entrez Gene ID: (Human) 415116, (Mouse) 223775
Molecular Function:
kinase
protein kinase
protein kinase receptor
receptor
serine/threonine protein kinase receptor
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.