OriGene
Manufacturer Code:TA501064
Catalog # 50-167-2207
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI10A12 |
Immunogen | Full length human recombit protein of human PIM2 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: pim-2 oncogene; Pim-2 proto-oncogene serine/threonine kinase; Pim-2h; proto-oncogene Pim-2 (serine threonine kinase); serine/threonine protein kinase pim-2; Serine/threonine-protein kinase pim-2
Gene Aliases: PIM2
UniProt ID: (Human) Q9P1W9
Entrez Gene ID: (Human) 11040
Molecular Function: kinase protein kinase protein kinase receptor receptor serine/threonine protein kinase receptor transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.