Invitrogen
Manufacturer Code:710504
Catalog # PI710504
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Oligoclonal |
Type | Antibody |
Clone | 19HCLC |
Immunogen | Peptide corresponding to amino acids 115130 of human PIM-1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Polyclonal Antibody
For Research Use Only
Protein Aliases: Oncogene PIM1; Oncogene PIM1 pim-1 kinase 44 kDa isoform pim-1 oncogene (proviral integration site 1) proto-oncogene serine/threonine-protein kinase pim-1; pim-1 kinase 44 kDa isoform; pim-1 oncogene; pim-1 oncogene (proviral integration site 1); proto-oncogene serine/threonine-protein kinase pim-1; Serine/threonine-protein kinase pim-1
Gene Aliases: PIM; Pim-1; PIM1
UniProt ID: (Human) P11309, (Mouse) P06803
Entrez Gene ID: (Human) 5292, (Mouse) 18712
Molecular Function: kinase protein kinase protein kinase receptor receptor serine/threonine protein kinase receptor transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.