Invitrogen
Manufacturer Code:PA536392
Catalog # PIPA536392
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 271-320 of human Pim-1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: non-specific serine/threonine protein kinase; Oncogene PIM1; pim-1 kinase 44 kDa isoform; pim-1 oncogene; pim-1 oncogene (proviral integration site 1); proto-oncogene serine/threonine-protein kinase Pim-1; proviral integration site 1; Serine/threonine-protein kinase pim-1; Serine/threonine-protein kinase pim-1 Name: Pim1 Pim-1
Gene Aliases: PIM; Pim-1; PIM1
UniProt ID: (Human) P11309, (Rat) P26794, (Mouse) P06803
Entrez Gene ID: (Human) 5292, (Rat) 24649, (Mouse) 18712
Molecular Function: kinase protein kinase protein kinase receptor receptor serine/threonine protein kinase receptor transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.