Novus Biologicals
Manufacturer Code:NBP231366
Catalog # NBP231366
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Partial recombinant protein made to an N-terminal portion of human PIM1 (between residues 1-200) [UniProt# P11309] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.11.1 Oncogene PIM1 PIM pim-1 kinase 44 kDa isoform pim-1 oncogene pim-1 oncogene (proviral integration site 1) proto-oncogene serine/threonine-protein kinase pim-1; Oncogene PIM1; pim-1 kinase 44 kDa isoform; pim-1 oncogene (proviral integration site 1); proto-oncogene serine/threonine-protein kinase pim-1; Serine/threonine-protein kinase pim-1
Gene Aliases: PIM; PIM1
UniProt ID: (Human) P11309
Entrez Gene ID: (Human) 5292
Molecular Function: kinase protein kinase protein kinase receptor receptor serine/threonine protein kinase receptor transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.