Novus Biologicals
Manufacturer Code:NBP273407H
Catalog # NBP273407H
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5D1 |
Immunogen | Recombinant protein fragment corresponding to amino acids 732-1036 of human PIK3R4 (NP_055417) produced in E.coli. |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: EC 2.7.11.1 MGC102700 p150 phosphatidylinositol 3-kinase-associated p150 Phosphoinositide 3-kinase adaptor protein phosphoinositide 3-kinase regulatory subunit 4 phosphoinositide-3-kinase regulatory subunit 4 phosphoinositide-3-kinase regulatory subunit 4 p150 PI3-kinase p150 subunit PI3-kinase regulatory subunit 4 VPS 15 VPS15; phosphatidylinositol 3-kinase-associated p150; Phosphoinositide 3-kinase adaptor protein; Phosphoinositide 3-kinase regulatory subunit 4; phosphoinositide-3-kinase, regulatory subunit 4; phosphoinositide-3-kinase, regulatory subunit 4, p150; PI3-kinase p150 subunit; PI3-kinase regulatory subunit 4
Gene Aliases: p150; PIK3R4; VPS15
UniProt ID: (Human) Q99570
Entrez Gene ID: (Human) 30849
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.