Novus Biologicals
Manufacturer Code:NBP130463
Catalog # NBP130463
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this product maps to a region between residue 1300 and 1358 of human vacuolar protein sorting 15 using the numbering given in entry NP_055417.1 (GeneID 30849). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.11.1 MGC102700 p150 phosphatidylinositol 3-kinase-associated p150 Phosphoinositide 3-kinase adaptor protein phosphoinositide 3-kinase regulatory subunit 4 phosphoinositide-3-kinase regulatory subunit 4 phosphoinositide-3-kinase regulatory subunit 4 p150 PI3-kinase p150 subunit PI3-kinase regulatory subunit 4 VPS 15 VPS15; phosphatidylinositol 3-kinase-associated p150; Phosphoinositide 3-kinase adaptor protein; Phosphoinositide 3-kinase regulatory subunit 4; phosphoinositide-3-kinase, regulatory subunit 4; phosphoinositide-3-kinase, regulatory subunit 4, p150; PI3-kinase p150 subunit; PI3-kinase regulatory subunit 4
Gene Aliases: p150; PIK3R4; VPS15
UniProt ID: (Human) Q99570
Entrez Gene ID: (Human) 30849
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.