OriGene
Manufacturer Code:CF801842
Catalog # 50-166-5745
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1F8 |
Immunogen | Human recombit protein fragment corresponding to amino acids 286-610 of human PIK3CD produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: APDS IMD14 P110DELTA PI3K p110D; Phosphatidylinositol 4,5-bisphosphate 3-kinase 110 kDa catalytic subunit delta; Phosphatidylinositol 4,5-bisphosphate 3-kinase catalytic subunit delta isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit delta; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit delta isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase, catalytic subunit delta; phosphoinositide-3-kinase C; phosphoinositide-3-kinase, catalytic, delta polypeptide variant p37delta; PI3-kinase p110 subunit delta; PI3-kinase subunit delta; PI3Kdelta; PtdIns-3-kinase subunit p110-delta
Gene Aliases: APDS; IMD14; p110D; P110DELTA; PI3K; PIK3CD
UniProt ID: (Human) O00329
Entrez Gene ID: (Human) 5293
Molecular Function: kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.