Invitrogen
Manufacturer Code:PA5115758
Catalog # PIPA5115758
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PIK3CD corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2410099E07Rik 2610208K16Rik APDS AW545373 catalytic phosphatidylinositol 3-kinase delta IMD14 p100 delta p110D P110DELTA phosphatidylinositol 3-kinase catalytic delta polypeptide phosphatidylinositol 3-kinase catalytic subunit delta phosphatidylinositol 3-kinase p110 delta phosphatidylinositol 45-bisphosphate 3-kinase 110 kDa catalytic subunit delta Phosphatidylinositol 45-bisphosphate 3-kinase catalytic subunit delta isoform phosphatidylinositol-45-bisphosphate 3-kinase 110 kDa catalytic subunit delta phosphatidylinositol-45-bisphosphate 3-kinase catalytic subunit delta phosphatidylinositol-45-bisphosphate 3-kinase catalytic subunit delta isoform phosphatidylinositol-45-bisphosphate 3-kinase catalytic subunit delta phosphoinositide 3 kinase P110delta phosphoinositide-3-kinase C phosphoinositide-3-kinase catalytic delta polypeptide phosphoinositide-3-kinase catalytic delta polypeptide variant p37delta PI3K PI3K delta pi3kcd PI3Kdelta PI3K-delta PI3-kinase p110 subunit delta PI3-kinase subunit delta PI3Kp110delta PI3K-p110delta PIK3CD ptdIns-3-kinase subunit delta PtdIns-3-kinase subunit p110-delta; catalytic phosphatidylinositol 3-kinase delta; p100 delta; phosphatidylinositol 3-kinase catalytic delta polypeptide; phosphatidylinositol 3-kinase catalytic subunit delta; phosphatidylinositol 3-kinase p110 delta; Phosphatidylinositol 4,5-bisphosphate 3-kinase 110 kDa catalytic subunit delta; Phosphatidylinositol 4,5-bisphosphate 3-kinase catalytic subunit delta isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit delta; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit delta isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase, catalytic subunit delta; phosphoinositide 3 kinase P110delta; phosphoinositide-3-kinase C; phosphoinositide-3-kinase, catalytic, delta polypeptide; phosphoinositide-3-kinase, catalytic, delta polypeptide variant p37delta; PI3-kinase p110 subunit delta; PI3-kinase subunit delta; PI3K-delta; PI3Kdelta; ptdIns-3-kinase subunit delta; PtdIns-3-kinase subunit p110-delta
Gene Aliases: 2410099E07Rik; 2610208K16Rik; APDS; AW545373; IMD14; p110D; P110DELTA; PI3K; PIK3CD
UniProt ID: (Human) O00329, (Mouse) O35904
Entrez Gene ID: (Human) 5293, (Mouse) 18707, (Rat) 366508
Molecular Function:
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.