Invitrogen
Manufacturer Code:MA514870
Catalog # PIMA514870
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | H.843.0 |
Immunogen | Synthetic peptide surrounding Asp520 of the sequence of human PI3K p110a |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: p110alpha; phosphatidylinositol 3-kinase, catalytic, 110-KD, alpha; phosphatidylinositol 3-kinase, catalytic, alpha polypeptide; Phosphatidylinositol 4,5-bisphosphate 3-kinase 110 kDa catalytic subunit alpha; Phosphatidylinositol 4,5-bisphosphate 3-kinase catalytic subunit alpha isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit alpha; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit alpha isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit, alpha isoform; Phosphoinositide 3-kinase alpha; Phosphoinositide-3-kinase catalytic alpha polypeptide; phosphoinositide-3-kinase, catalytic, alpha polypeptide; PI3-kinase p110 subunit alpha; PI3-kinase p110 subunit alpha PI3-kinase subunit alpha PI3K-alpha PtdIns-3-kinase p110 phosphatidylinositol 3-kinase catalytic 110-KD alpha phosphatidylinositol 3-kinase catalytic alpha polypeptide phosphatidylinositol-45-bisphosphate 3-kinase 110 kDa catalytic subunit alpha phosphatidylinositol-45-bisphosphate 3-kinase catalytic subunit alpha isoform phosphatidylinositol-45-bisphosphate 3-kinase catalytic subunit alpha isoform ptdIns-3-kinase subunit alpha ptdIns-3-kinase sub; PI3-kinase subunit alpha; PI3K; PI3K-alpha; PI3Kalpha; ptdIns-3-kinase p110; ptdIns-3-kinase subunit alpha; PtdIns-3-kinase subunit p110-alpha; Serine/threonine protein kinase PIK3CA
Gene Aliases: 6330412C24Rik; caPI3K; CLOVE; CWS5; MCAP; MCM; MCMTC; p110; p110-alpha; p110alpha; PI3K; PI3K-alpha; PIK3CA
UniProt ID: (Bovine) P32871, (Human) P42336, (Mouse) P42337, (Rat) A0A0G2K344
Entrez Gene ID: (Bovine) 282306, (Human) 5290, (Mouse) 18706, (Rat) 170911
Molecular Function:
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.