Novus Biologicals
Manufacturer Code:NBP219804
Catalog # NBP219804
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the N-terminus region of human PI3 kinase p110 alpha. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.1 EC 2.7.1.153 MGC142161 MGC142163 p110-alpha Phosphatidylinositol 3-Kinase catalytic subunit alpha phosphatidylinositol 3-kinase catalytic 110-KD alpha phosphatidylinositol 3-kinase catalytic alpha polypeptide Phosphatidylinositol-45-bisphosphate 3-kinase 110 kDa catalytic subunit alpha phosphatidylinositol-45-bisphosphate 3-kinase catalytic subunit alpha isoform phosphatidylinositol-45-bisphosphate 3-kinase catalytic subunit alpha isoform phosphoinositide-3-kinase catalytic alpha polypeptide PI3K PI3K-alpha PI3-kinase p110 subunit alpha PI3-kinase subunit alpha PtdIns-3-kinase p110 PtdIns-3-kinase subunit alpha PtdIns-3-kinase subunit p110-alpha; phosphatidylinositol 3-kinase, catalytic, 110-KD, alpha; phosphatidylinositol 3-kinase, catalytic, alpha polypeptide; Phosphatidylinositol 4,5-bisphosphate 3-kinase 110 kDa catalytic subunit alpha; Phosphatidylinositol 4,5-bisphosphate 3-kinase catalytic subunit alpha isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit alpha; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit, alpha isoform; Phosphoinositide 3-kinase alpha; Phosphoinositide-3-kinase catalytic alpha polypeptide; phosphoinositide-3-kinase, catalytic, alpha polypeptide; PI3-kinase p110 subunit alpha; PI3-kinase subunit alpha; PtdIns-3-kinase subunit p110-alpha; Serine/threonine protein kinase PIK3CA
Gene Aliases: CLOVE; CWS5; MCAP; MCM; MCMTC; p110-alpha; PI3K; PI3K-alpha; PIK3CA
UniProt ID: (Human) P42336
Entrez Gene ID: (Human) 5290
Molecular Function: kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.