Invitrogen
Manufacturer Code:PA575656
Catalog # PIPA575656
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues in Human PIK3C2G. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: phosphatidylinositol 3-kinase, C2 domain containing, gamma polypeptide; Phosphatidylinositol 4-phosphate 3-kinase C2 domain-containing subunit gamma; Phosphatidylinositol 4-phosphate 3-kinase C2 domain-containing subunit gamma PI3K-C2-gamma Short name&equalsPtdIns-3-kinase C2 subunit gamma Phosphoinositide 3-kinase-C2-gamma PI 3-kinase C2gamma PI3KC2gamma; phosphatidylinositol-4-phosphate 3-kinase C2 domain-containing gamma polypeptide; phosphatidylinositol-4-phosphate 3-kinase C2 domain-containing subunit gamma; Phosphoinositide 3-kinase-C2-gamma; phosphoinositide-3-kinase, class 2, gamma polypeptide; PI3K-C2-gamma; PI3K-C2gamma; PTDINS-3-kinase C2 gamma; ptdIns-3-kinase C2 subunit gamma
Gene Aliases: C80387; PI3K-C2-gamma; PI3K-C2GAMMA; PIK3C2G
UniProt ID: (Human) O75747, (Rat) O70173, (Mouse) O70167
Entrez Gene ID: (Human) 5288, (Rat) 116720, (Mouse) 18705
Molecular Function: kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.