Invitrogen
Manufacturer Code:PA5100895
Catalog # PIPA5100895
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PIK3C2B corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C2-PI3K; C2-PI3K C330011J12Rik Gm199 phosphatidylinositol 3-kinase C2 domain-containing beta polypeptide phosphatidylinositol 4-phosphate 3-kinase C2 domain-containing subunit beta phosphatidylinositol-4-phosphate 3-kinase C2 domain-containing subunit beta phosphatidylinositol-4-phosphate 3-kinase catalytic subunit type 2 beta phosphatidylinositol-4-phosphate 3-kinase catalytic subunit type 2 beta phosphoinositide 3-kinase-C2-beta phosphoinositide-3-kinase class 2 beta polypeptide pi3kc2b PI3KC2beta PI3K-C2beta PI3K-C2-beta PIK3C2B PTDINS-3-kinase C2 beta ptdIns-3-kinase C2 subunit beta; phosphatidylinositol 3-kinase C2 domain-containing beta polypeptide; Phosphatidylinositol 4-phosphate 3-kinase C2 domain-containing subunit beta; phosphatidylinositol-4-phosphate 3-kinase C2 domain-containing subunit beta; phosphatidylinositol-4-phosphate 3-kinase, catalytic subunit type 2 beta; Phosphoinositide 3-kinase-C2-beta; phosphoinositide-3-kinase, class 2, beta polypeptide; PI3K-C2-beta; PI3K-C2beta; PTDINS-3-kinase C2 beta; ptdIns-3-kinase C2 subunit beta
Gene Aliases: C2-PI3K; C330011J12Rik; Gm199; PI3K-C2beta; PI3KC2beta; PIK3C2B
UniProt ID: (Human) O00750
Entrez Gene ID: (Human) 5287, (Mouse) 240752
Molecular Function:
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.