Novus Biologicals
Manufacturer Code:NBP298034AF700
Catalog # NB051378
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human PIH1D3 fragment. |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: chromosome X open reading frame 41 CXorf41 hypothetical protein LOC139212 MGC35261 NYSAR97 PIH1 domain containing 3 Sarcoma antigen NY-SAR-97; Dynein assembly factor 6, axonemal; PIH1 domain-containing protein 3; Sarcoma antigen NY-SAR-97
Gene Aliases: CXorf41; DNAAF6; NYSAR97; PIH1D3
UniProt ID: (Human) Q9NQM4
Entrez Gene ID: (Human) 139212
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.