Invitrogen
Manufacturer Code:PA563595
Catalog # PIPA563595
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human PIGZ |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dol-P-Man dependent GPI mannosyltransferase; GPI mannosyltransferase 4; GPI mannosyltransferase IV; GPI-MT-IV; GPI-MT-IV PIG-Z SMP3 GPI mannosyltransferase 4 GPI mannosyltransferase IV SMP3 mannosyltransferase phosphatidylinositol glycan class Z phosphatidylinositol-glycan biosynthesis class Z protein; hSMP3; phosphatidylinositol glycan anchor biosynthesis, class Z; phosphatidylinositol glycan, class Z; Phosphatidylinositol-glycan biosynthesis class Z protein; PIG-Z; SMP3 homolog; SMP3 mannosyltransferase
Gene Aliases: GPI-MT-IV; PIG-Z; PIGZ; SMP3
UniProt ID: (Human) Q86VD9
Entrez Gene ID: (Human) 80235
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.