Proteintech
Manufacturer Code:173841AP150UL
Catalog # 50-173-1542
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PIGZ Fusion Protein Ag11259 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dol-P-Man dependent GPI mannosyltransferase; GPI mannosyltransferase 4; GPI mannosyltransferase 4 GPI mannosyltransferase IV GPI MT IV hSMP3 PIG Z PIGZ SMP3 SMP3 homolog; GPI mannosyltransferase IV; GPI-MT-IV; hSMP3; phosphatidylinositol glycan anchor biosynthesis, class Z; phosphatidylinositol glycan, class Z; phosphatidylinositol glycan, class Z-like 1; Phosphatidylinositol-glycan biosynthesis class Z protein; PIG-Z; SMP3 homolog; SMP3 mannosyltransferase
Gene Aliases: F630022B06Rik; GPI-MT-IV; PIG-Z; PIGZ; RGD1307107; SMP3
UniProt ID: (Human) Q86VD9, (Mouse) Q8BTP0
Entrez Gene ID: (Human) 80235, (Mouse) 239827, (Rat) 303872
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.