Invitrogen
Manufacturer Code:PA556134
Catalog # PIPA556134
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human PIGX |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GPI-mannosyltransferase subunit; GPI-mannosyltransferase subunit PIG-X phosphatidylinositol-glycan biosynthesis class X protein; phosphatidylinositol glycan anchor biosynthesis, class X; Phosphatidylinositol-glycan biosynthesis class X protein; PIG-X
Gene Aliases: PIG-X; PIGX
UniProt ID: (Human) Q8TBF5
Entrez Gene ID: (Human) 54965
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.