Invitrogen
Manufacturer Code:PA5113251
Catalog # PIPA5113251
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Phosphatidylinositol-glycan biosynthesis class X protein (42-230AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GPI-mannosyltransferase subunit; GPI-mannosyltransferase subunit phosphatidylinositol glycan anchor biosynthesis class X phosphatidylinositol glycan anchor biosynthesis class X Phosphatidylinositol-glycan biosynthesis class X protein PIGX PIG-X; phosphatidylinositol glycan anchor biosynthesis, class X; phosphatidylinositol glycan, class X; Phosphatidylinositol-glycan biosynthesis class X protein; PIG-X
Gene Aliases: 2010319C14Rik; PIG-X; PIGX
UniProt ID: (Human) Q8TBF5, (Mouse) Q99LV7
Entrez Gene ID: (Human) 54965, (Mouse) 72084
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.