Invitrogen
Manufacturer Code:PA5113078
Catalog # PIPA5113078
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Phosphatidylinositol-glycan biosynthesis class W protein (392-448AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2610044A17Rik Gwt1 HPMRS5 phosphatidylinositol glycan anchor biosynthesis class W phosphatidylinositol glycan anchor biosynthesis class W phosphatidylinositol glycan class W Phosphatidylinositol-glycan biosynthesis class W protein PIGW PIG-W; phosphatidylinositol glycan anchor biosynthesis, class W; phosphatidylinositol glycan, class W; Phosphatidylinositol-glycan biosynthesis class W protein; PIG-W
Gene Aliases: Gwt1; HPMRS5; PIGW
UniProt ID: (Human) Q7Z7B1, (Rat) Q7TSN4
Entrez Gene ID: (Human) 284098, (Rat) 378774
Molecular Function: acetyltransferase acyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.