Novus Biologicals
Manufacturer Code:NBP160017
Catalog # NBP160017
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to PIGW(phosphatidylinositol glycan anchor biosynthesis class W) The peptide sequence was selected from the middle region of PIGW. Peptide sequence: IALGITVLYQLALDFTSLKRLILYGTDGSGTRVGLLNANREGIISTLGYV. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: class W EC 2.3 FLJ37433 phosphatidylinositol glycan anchor biosynthesis class W PIG-W; phosphatidylinositol glycan anchor biosynthesis, class W; phosphatidylinositol glycan, class W; Phosphatidylinositol-glycan biosynthesis class W protein; PIG-W
Gene Aliases: Gwt1; HPMRS5; PIGW
UniProt ID: (Human) Q7Z7B1
Entrez Gene ID: (Human) 284098
Molecular Function: acetyltransferase acyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.