Invitrogen
Manufacturer Code:PA543022
Catalog # PIPA543022
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human PIGV |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dol-P-Man dependent GPI mannosyltransferase; GPI mannosyltransferase 2; GPI mannosyltransferase II; GPI-MT-II; Phosphatidylinositol glycan anchor biosynthesis class V FLJ20477 PIG-V GPI-MT-II; phosphatidylinositol glycan anchor biosynthesis, class V; phosphatidylinositol glycan class V; phosphatidylinositol glycan, class V; Phosphatidylinositol-glycan biosynthesis class V protein; PIG-V; Ybr004c homolog
Gene Aliases: B330013B03; D430024F16Rik; GPI-MT-II; HPMRS1; PIG-V; PIGV; RGD1309526
UniProt ID: (Human) Q9NUD9, (Rat) Q5KR61, (Mouse) Q7TPN3
Entrez Gene ID: (Rabbit) 100358444, (Human) 55650, (Bovine) 519213, (Dog) 106557463, (Rat) 366478, (Guinea pig) 100727680, (Mouse) 230801, (Zebrafish) 100148642
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.