Invitrogen
Manufacturer Code:PA576848
Catalog # PIPA576848
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human PIGR. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hepatocellular carcinoma associated protein TB6; Hepatocellular carcinoma associated protein TB6 Hepatocellular carcinoma-associated protein TB6 Phosphatidylinositol glycan class R PIGR PIGR&lowbarHUMAN Poly Ig receptor Poly-Ig receptor Polymeric immunoglobulin receptor Secretory component Transmembrane secretory component; Hepatocellular carcinoma-associated protein TB6; phosphatidylinositol glycan, class R; PIgR; poly-Ig receptor; Polymeric immunoglobulin receptor; Secretory component
Gene Aliases: PIGR
UniProt ID: (Human) P01833, (Mouse) O70570
Entrez Gene ID: (Human) 5284, (Mouse) 18703
Molecular Function:
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.