Invitrogen
Manufacturer Code:MA524423
Catalog # PIMA524423
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2B7 |
Immunogen | PIGQ (NP&lowbar683721.1 661 a.a. &bsim 758 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C,Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: c407A10.1 (GPI1 (N-acetylglucosaminyl transferase component)); N-acetylglucosaminyl transferase component Gpi1; N-acetylglucosamyl transferase component GPI1; phosphatidylinositol glycan anchor biosynthesis, class Q; phosphatidylinositol glycan, class Q; Phosphatidylinositol N-acetylglucosaminyltransferase subunit Q; Phosphatidylinositol-glycan biosynthesis class Q protein; PIG-Q; PIGQ GPI1
Gene Aliases: c407A10.1; GPI1; PIGQ
UniProt ID: (Human) Q9BRB3
Entrez Gene ID: (Human) 9091
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.