Invitrogen
Manufacturer Code:PA5104009
Catalog # PIPA5104009
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PIGQ corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: c407A10.1 (GPI1 (N-acetylglucosaminyl transferase component)); c407A10.1 c407A10.1 (GPI1 (N-acetylglucosaminyl transferase component)) glycosylphosphatidylinositol 1 homolog GPI1 Gpi1h Gpi1p Gpih Mgpi1 MGpi1p N-acetylglucosaminyl transferase component Gpi1 N-acetylglucosamyl transferase component GPI1 phosphatidylinositol glycan anchor biosynthesis class Q phosphatidylinositol glycan anchor biosynthesis class Q phosphatidylinositol glycan class Q Phosphatidylinositol N-acetylglucosaminyltransferase subunit Q phosphatidylinositol-glycan biosynthesis class Q protein Pigq PIG-Q; glycosylphosphatidylinositol 1 homolog; MGpi1p; N-acetylglucosaminyl transferase component Gpi1; N-acetylglucosamyl transferase component GPI1; phosphatidylinositol glycan anchor biosynthesis, class Q; phosphatidylinositol glycan, class Q; Phosphatidylinositol N-acetylglucosaminyltransferase subunit Q; Phosphatidylinositol-glycan biosynthesis class Q protein; PIG-Q
Gene Aliases: c407A10.1; GPI1; Gpi1h; Gpi1p; Gpih; Mgpi1; PIGQ
UniProt ID: (Human) Q9BRB3, (Mouse) Q9QYT7
Entrez Gene ID: (Human) 9091, (Rat) 287159, (Mouse) 14755
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.