Invitrogen
Manufacturer Code:PA5104008
Catalog # PIPA5104008
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PIGP corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DCRC DCRC-S Down syndrome critical region gene 5 Down syndrome critical region gene c Down syndrome critical region homolog 5 Down syndrome critical region protein 5 Down syndrome critical region protein 5 homolog Down syndrome critical region protein C DSCR5 DSCRC DSRC NPD010 phosphatidylinositol glycan anchor biosynthesis class P phosphatidylinositol glycan anchor biosynthesis class P phosphatidylinositol glycan class P phosphatidylinositol N-acetylglucosaminyltransferase subunit P Phosphatidylinositol-glycan biosynthesis class P protein phosphatidylinositol-n-acetylglucosaminyltranferase subunit PIGP PIG-P; Down syndrome critical region gene 5; Down syndrome critical region gene c; Down syndrome critical region homolog 5; Down syndrome critical region protein 5; Down syndrome critical region protein 5 homolog; Down syndrome critical region protein C; phosphatidylinositol glycan anchor biosynthesis, class P; phosphatidylinositol glycan, class P; Phosphatidylinositol N-acetylglucosaminyltransferase subunit P; Phosphatidylinositol-glycan biosynthesis class P protein; phosphatidylinositol-n-acetylglucosaminyltranferase subunit; PIG-P
Gene Aliases: DCRC; DCRC-S; DSCR5; DSCRC; DSRC; NPD010; PIG-P; PIGP
UniProt ID: (Human) Q53Y28, (Mouse) Q9JHG1
Entrez Gene ID: (Human) 51227, (Mouse) 56176
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.