Novus Biologicals
Manufacturer Code:NBP294792100UL
Catalog # NB024097
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 25-158 of human PIGP (NP_710148.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: class P DCRCDown syndrome critical region protein C Down syndrome critical region protein 5 DSCR5DCRC-S DSCRC DSRC EC 2.4.1.198 phosphatidylinositol glycan anchor biosynthesis class P Phosphatidylinositol-glycan biosynthesis class P protein phosphatidylinositol-n-acetylglucosaminyltranferase subunit PIG-P; Down syndrome critical region gene 5; Down syndrome critical region protein 5; Down syndrome critical region protein C; phosphatidylinositol glycan anchor biosynthesis, class P; phosphatidylinositol glycan, class P; Phosphatidylinositol N-acetylglucosaminyltransferase subunit P; Phosphatidylinositol-glycan biosynthesis class P protein; phosphatidylinositol-n-acetylglucosaminyltranferase subunit; PIG-P
Gene Aliases: DCRC; DCRC-S; DSCR5; DSCRC; DSRC; NPD010; PIG-P; PIGP
UniProt ID: (Human) Q53Y28
Entrez Gene ID: (Human) 51227
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.