Proteintech
Manufacturer Code:163691AP150UL
Catalog # 50-173-0897
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PIGO Fusion Protein Ag9232 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GPI ethanolamine phosphate transferase 3; phosphatidylinositol glycan anchor biosynthesis, class O; phosphatidylinositol glycan, class O; Phosphatidylinositol-glycan biosynthesis class O protein; PIG O PIGO RP11 182N22.4; PIG-O
Gene Aliases: HPMRS2; PIG-O; PIGO; UNQ632/PRO1249
UniProt ID: (Human) Q8TEQ8
Entrez Gene ID: (Human) 84720, (Mouse) 56703, (Rat) 313341
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.