Abnova
Manufacturer Code:PAB23600
Catalog # 89-125-306
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human PIGN. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant PIGN.
For Research Use Only
Protein Aliases: GPI ethanolamine phosphate transferase 1; MCD4; MCD4 homolog; MDC4; MGC26427; phosphatidylinositol glycan anchor biosynthesis, class N; Phosphatidylinositol-glycan biosynthesis class N protein; PIG-N
Gene Aliases: MCAHS; MCAHS1; MCD4; MDC4; PIG-N; PIGN
UniProt ID: (Human) O95427
Entrez Gene ID: (Human) 23556
Molecular Function:
extracellular matrix glycoprotein
extracellular matrix protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.