Proteintech
Manufacturer Code:204211AP150UL
Catalog # 50-173-2880
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PIGM Fusion Protein Ag14245 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dol-P-Man dependent GPI mannosyltransferase; DPM:GlcN-(acyl-)PI mannosyltransferase; GPI mannosyltransferase 1; GPI mannosyltransferase 1 GPI mannosyltransferase I GPI MT I PIG M PIGM; GPI mannosyltransferase I; GPI-MT-I; phosphatidylinositol glycan anchor biosynthesis, class M; phosphatidylinositol glycan, class M; Phosphatidylinositol-glycan biosynthesis class M protein; PIG-M; PIG-M mannosyltransferase; PIG-M mRNA for mannosyltransferase
Gene Aliases: 4933437L05Rik; AI644422; AV302080; C920011G20Rik; GPI-MT-I; PIGM
UniProt ID: (Human) Q9H3S5, (Mouse) Q8C2R7, (Rat) Q9EQY6
Entrez Gene ID: (Human) 93183, (Mouse) 67556, (Rat) 79112
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.