Invitrogen
Manufacturer Code:PA528337
Catalog # PIPA528337
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 314 of Human PIGK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GPI transamidase; GPI transamidase subunit; GPI-anchor transamidase; GPI8 GPI transamidase subunit GPI-anchor transamidase GPI8 homolog PIG-K phosphatidylinositol glycan class K phosphatidylinositol-glycan biosynthesis class K protein; GPI8 homolog; hGPI8; phosphatidylinositol glycan anchor biosynthesis, class K; phosphatidylinositol glycan, class K; Phosphatidylinositol-glycan biosynthesis class K protein; PIG-K
Gene Aliases: 3000001O05Rik; GPI8; PIGK
UniProt ID: (Human) Q92643, (Mouse) Q9CXY9
Entrez Gene ID: (Human) 10026, (Mouse) 329777
Molecular Function:
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.