Invitrogen
Manufacturer Code:PA553409
Catalog # PIPA553409
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human PIGG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GPI ethanolamine phosphate transferase 2; GPI7 homolog; hGPI7; phosphatidylinositol glycan anchor biosynthesis, class G; Phosphatidylinositol-glycan biosynthesis class G protein; PIG-G
Gene Aliases: GPI7; LAS21; MRT53; PIGG; PRO4405; RLGS1930; UNQ1930/PRO4405
UniProt ID: (Human) Q5H8A4
Entrez Gene ID: (Human) 54872
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.