Abnova
Manufacturer Code:PAB20864
Catalog # 89-122-362
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human PIGG. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant PIGG.
For Research Use Only
Protein Aliases: DKFZp434A1810; DKFZp434C1712; DKFZp434M1131; FLJ20265; FLJ39925; GPI ethanolamine phosphate transferase 2; GPI7; GPI7 homolog; hGPI7; LAS21; MGC131903; phosphatidylinositol glycan anchor biosynthesis, class G; Phosphatidylinositol-glycan biosynthesis class G protein; PIG-G; PRO4405; RLGS1930
Gene Aliases: GPI7; LAS21; MRT53; PIGG; PRO4405; RLGS1930; UNQ1930/PRO4405
UniProt ID: (Human) Q5H8A4
Entrez Gene ID: (Human) 54872
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.