Novus Biologicals
Manufacturer Code:NB11085468C
Catalog # NB11085468C
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | RM0010-8F09 |
Immunogen | Mouse recombinant PlGF protein. |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rat Monoclonal Antibody
Protein Aliases: GPI11 homolog; GPI11 homolog MGC32646 MGC33136 phosphatidylinositol glycan anchor biosynthesis class F phosphatidylinositol glycan class F phosphatidylinositol-glycan biosynthesis class F protein PIG-F; phosphatidylinositol glycan anchor biosynthesis, class F; Phosphatidylinositol-glycan biosynthesis class F protein; PIG-F
Gene Aliases: PIGF
UniProt ID: (Human) Q07326
Entrez Gene ID: (Human) 5281
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.