Invitrogen
Manufacturer Code:PA575899
Catalog # PIPA575899
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues in Human PIGF. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GPI11 homolog; phosphatidylinositol glycan anchor biosynthesis, class F; phosphatidylinositol glycan, class F; Phosphatidylinositol-glycan biosynthesis class F protein; Phosphatidylinositol-glycan biosynthesis class F protein PIG-F GPI11 homolog PIGF; PIG-F
Gene Aliases: PIGF
UniProt ID: (Human) Q07326, (Mouse) O09101
Entrez Gene ID: (Human) 5281, (Rat) 681086, (Mouse) 18701
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.