Invitrogen
Manufacturer Code:PA5104007
Catalog # PIPA5104007
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PIGC corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2.4.1.198 3110030E07Rik AW212108 GPI2 phosphatidylinositol glycan anchor biosynthesis class C phosphatidylinositol glycan anchor biosynthesis class C phosphatidylinositol glycan class C phosphatidylinositol N-acetylglucosaminyltransferase subunit C Phosphatidylinositol-glycan biosynthesis class C protein phosphatidylinositol-glycan biosynthesis class C protein Pigc PIG-C; phosphatidylinositol glycan, class C; Phosphatidylinositol N-acetylglucosaminyltransferase subunit C; Phosphatidylinositol-glycan biosynthesis class C protein; phosphatidylinositol-glycan biosynthesis, class C protein; PIG-C
Gene Aliases: 3110030E07Rik; AW212108; GPI2; PIGC
UniProt ID: (Human) Q92535, (Mouse) Q9CXR4, (Rat) Q5PQQ4
Entrez Gene ID: (Human) 5279, (Mouse) 67292, (Rat) 364032
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.