Novus Biologicals
Manufacturer Code:NBP17987120UL
Catalog # NBP1798720
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C terminal of human PIGCThe immunogen for this antibody is PIGC. Peptide sequence AVGAVLFALLLMSISCLCPFYLIRLQLFKENIHGPWDEAEIKEDLSRFLS. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.4.1.198 GPI2class C protein MGC2049 phosphatidylinositol glycan anchor biosynthesis class C phosphatidylinositol glycan class C; Phosphatidylinositol N-acetylglucosaminyltransferase subunit C; Phosphatidylinositol-glycan biosynthesis class C protein; phosphatidylinositol-glycan biosynthesis, class C protein; PIG-C
Gene Aliases: GPI2; PIGC
UniProt ID: (Human) Q92535
Entrez Gene ID: (Human) 5279
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.