Invitrogen
Manufacturer Code:MA524388
Catalog # PIMA524388
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2G7 |
Immunogen | PIGB (NP&lowbar004846 80 a.a. &bsim 172 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C,Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: dol-P-Man dependent GPI mannosyltransferase; GPI mannosyltransferase 3; GPI mannosyltransferase 3 241- GPI mannosyltransferase III GPI-MT-III Phosphatidylinositol-glycan biosynthesis class B protein PIG-B PIGB; GPI mannosyltransferase III; GPI-MT-III; phosphatidylinositol glycan anchor biosynthesis, class B; phosphatidylinositol glycan, class B; Phosphatidylinositol-glycan biosynthesis class B protein; PIG-B
Gene Aliases: GPI-MT-III; PIG-B; PIGB
UniProt ID: (Human) Q92521
Entrez Gene ID: (Human) 9488
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.