Invitrogen
Manufacturer Code:PA558993
Catalog # PIPA558993
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human PIGB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dol-P-Man dependent GPI mannosyltransferase; GPI mannosyltransferase 3; GPI mannosyltransferase 3 241- GPI mannosyltransferase III GPI-MT-III Phosphatidylinositol-glycan biosynthesis class B protein PIG-B PIGB; GPI mannosyltransferase III; GPI-MT-III; involved in GPI-anchor biosynthesis; phosphatidylinositol glycan anchor biosynthesis, class B; phosphatidylinositol glycan, class B; Phosphatidylinositol-glycan biosynthesis class B protein; PIG-B
Gene Aliases: GPI-MT-III; PIG-B; PIGB
UniProt ID: (Human) Q92521, (Mouse) Q9JJQ0
Entrez Gene ID: (Human) 9488, (Mouse) 55981, (Rat) 315807
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.