Invitrogen
Manufacturer Code:PA527740
Catalog # PIPA527740
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 270 and 484 of Human PIGA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GlcNAc-PI synthesis protein; GPI3 PIG-A GLCNAC-PI synthesis protein GPI anchor biosynthesis class A GlcNAc-inositol phospholipid assembly protein phosphatidylinositol N-acetylglucosaminyltransferase subunit A phosphatidylinositol-glycan biosynthesis class A protein; N-acetylglucosaminyl-phosphatidylinositol biosynthetic protein; phosphatidylinositol glycan, class A; Phosphatidylinositol N-acetylglucosaminyltransferase subunit A; Phosphatidylinositol-glycan biosynthesis class A protein; phosphatidylinositolglycan class A; PIG-A
Gene Aliases: AI194334; Pig-a; Piga
UniProt ID: (Mouse) Q64323
Entrez Gene ID: (Mouse) 18700
Molecular Function: carbohydrate phosphatase glycosyltransferase hydrolase nucleotidyltransferase phosphatase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.