Invitrogen
Manufacturer Code:PA5104006
Catalog # PIPA5104006
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PIGA corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI194334 class A GlcNAc-inositol phospholipid assembly protein glcNAc-PI synthesis protein GPI anchor biosynthesis GPI3 MCAHS2 N-acetylglucosaminyl-phosphatidylinositol biosynthetic protein phosphatidylinositol glycan anchor biosynthesis class A phosphatidylinositol glycan anchor biosynthesis class A phosphatidylinositol glycan class A phosphatidylinositol N-acetylglucosaminyltransferase subunit A Phosphatidylinositol-glycan biosynthesis class A protein phosphatidylinositol-glycan biosynthesis class A protein phosphatidylinositolglycan class A PIGA Pig-a PNH1; class A GlcNAc-inositol phospholipid assembly protein; GLCNAC-PI synthesis protein; GPI anchor biosynthesis; N-acetylglucosaminyl-phosphatidylinositol biosynthetic protein; phosphatidylinositol glycan anchor biosynthesis, class A; phosphatidylinositol glycan, class A; Phosphatidylinositol N-acetylglucosaminyltransferase subunit A; Phosphatidylinositol-glycan biosynthesis class A protein; phosphatidylinositol-glycan biosynthesis, class A protein; phosphatidylinositolglycan class A; PIG-A
Gene Aliases: AI194334; GPI3; MCAHS2; PIG-A; PIGA; PNH1
UniProt ID: (Human) P37287, (Mouse) Q64323
Entrez Gene ID: (Human) 5277, (Rat) 363464, (Mouse) 18700
Molecular Function: carbohydrate phosphatase glycosyltransferase hydrolase nucleotidyltransferase phosphatase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.