Invitrogen
Manufacturer Code:PA572989
Catalog # PIPA572989
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to an internal portion of the human PIF1 protein (between residues 400-500) &lsqbUniProt Q9H611] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent DNA helicase PIF1; ATP-dependent DNA helicase PIF1 DNA repair and recombination helicase PIF-1 PIF1/RRM3 DNA helicase-like protein C15orf20; DNA helicase-like protein; DNA repair and recombination helicase PIF1; PIF1 homolog; Pif1/Rrm3 DNA helicase-like protein
Gene Aliases: 4631410M14; AI449441; Pif1
UniProt ID: (Mouse) Q80SX8
Entrez Gene ID: (Mouse) 208084
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.