Invitrogen
Manufacturer Code:PA5103641
Catalog # PIPA5103641
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PIF1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4631410M14 AI449441 ATP-dependent DNA helicase PIF1 ATP-dependent DNA helicase PIF1 {ECO:0000255 C15orf20 DNA helicase-like protein DNA repair and recombination helicase PIF1 DNA repair and recombination helicase PIF1 {ECO:0000255 HAMAP-Rule:MF_03176} petite integration frequency 1 PIF PIF1 PIF1 5'-to-3' DNA helicase PIF1 5'-to-3' DNA helicase homolog PIF1 5'-to-3' DNA helicase homolog (S. cerevisiae) PIF1 homolog Pif1/Rrm3 DNA helicase-like protein Pif1/Rrm3 DNA-helicase-like protein; ATP-dependent DNA helicase PIF1; ATP-dependent DNA helicase PIF1 {ECO:0000255|HAMAP-Rule:MF_03176}; DNA helicase-like protein; DNA repair and recombination helicase PIF1; DNA repair and recombination helicase PIF1 {ECO:0000255|HAMAP-Rule:MF_03176}; petite integration frequency 1; PIF1 5'-to-3' DNA helicase homolog; PIF1 homolog; PIF1/RRM3 DNA helicase-like protein; Pif1/Rrm3 DNA-helicase-like protein
Gene Aliases: 4631410M14; AI449441; C15orf20; PIF; PIF1
UniProt ID: (Human) Q9H611, (Rat) Q1HG60, (Mouse) Q80SX8
Entrez Gene ID: (Human) 80119, (Rat) 367645, (Mouse) 208084
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.