Novus Biologicals
Manufacturer Code:NBP197931H
Catalog # NBP197931H
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to an internal portion of the human PIF1 protein (between residues 400-500) [UniProt Q9H611] |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent DNA helicase PIF1; C15orf20 PIF PIF1 5-to-3 Prime DNA helicase homolog RRM3 DNA helicase-like protein; DNA repair and recombination helicase PIF1; petite integration frequency 1; PIF1 5'-to-3' DNA helicase homolog; PIF1/RRM3 DNA helicase-like protein
Gene Aliases: C15orf20; PIF; PIF1
UniProt ID: (Human) Q9H611
Entrez Gene ID: (Human) 80119
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.