Novus Biologicals
Manufacturer Code:NBP29382020UL
Catalog # NB029501
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 124-248 of human PID1 (NP_060403.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HMFN2073 P-CLI1 phosphotyrosine interaction domain containing 1 PTB-containing cubilin and LRP1-interacting protein; P-CLI1; Phosphotyrosine interaction domain-containing protein 1; Protein NYGGF4; PTB-containing, cubilin and LRP1-interacting protein
Gene Aliases: HMFN2073; NYGGF4; P-CLI1; PCLI1; PID1
UniProt ID: (Human) Q7Z2X4
Entrez Gene ID: (Human) 55022
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.