Invitrogen
Manufacturer Code:PA513841
Catalog # PIPA513841
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 286-316 amino acids from the C-terminal region of human PICK1 (PRKCABP) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PRKCA-binding protein; PRKCA-binding protein Protein kinase C-alpha binding protein Protein interacting with C kinase 1 PICK1; Protein interacting with C kinase 1; Protein kinase C-alpha-binding protein; protein that interacts with C kinase 1
Gene Aliases: PICK; PICK1; PRKCABP
UniProt ID: (Human) Q9NRD5
Entrez Gene ID: (Human) 9463, (Mouse) 18693
Molecular Function: G-protein modulator enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.