Novus Biologicals
Manufacturer Code:NB10041403
Catalog # NB10041403
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-HYNKLQADPKQ corresponding to internal region according to NP_036539.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha binding protein dJ1039K5 MGC15204 PICK PRKCA-binding protein PRKCABPprotein interacting with PRKCA Protein interacting with C kinase 1 protein interacting with PRKCA 1 Protein kinase C-alpha-binding protein; PRKCA-binding protein; Protein interacting with C kinase 1; Protein kinase C-alpha-binding protein
Gene Aliases: PICK; PICK1; PRKCABP
UniProt ID: (Human) Q9NRD5
Entrez Gene ID: (Human) 9463
Molecular Function: G-protein modulator enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.