Novus Biologicals
Manufacturer Code:NB10058835
Catalog # NB10058835
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 735 and 757 of human progesterone-induced-blocking factor 1 using the numbering given in entry NP_006337.2 (GeneID 10464). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C13orf24 chromosome 13 open reading frame 24 KIAA1008 PIBF progesterone immunomodulatory binding factor 1 progesterone-induced blocking factor 1 progesterone-induced-blocking factor 1 RP11-505F3.1; Centrosomal protein of 90 kDa; CEP90; PIBF; progesterone-induced blocking factor 1; Progesterone-induced-blocking factor 1
Gene Aliases: C13orf24; CEP90; PIBF; PIBF1
UniProt ID: (Human) Q8WXW3
Entrez Gene ID: (Human) 10464
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.