Novus Biologicals
Manufacturer Code:NBP2823080.1ML
Catalog # NB030237
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of human PIB5PA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A inositol polyphosphate 5-phosphatase J inositol polyphosphate-5-phosphatase J phosphatidylinositol 45-bisphosphate 5-phosphatase A PIPP; Inositol polyphosphate 5-phosphatase J; phosphatidylinositol (4,5) bisphosphate 5-phosphatase, A; Phosphatidylinositol 4,5-bisphosphate 5-phosphatase A
Gene Aliases: INPP5; INPP5J; PIB5PA; PIPP
UniProt ID: (Human) Q15735
Entrez Gene ID: (Human) 27124
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.