Invitrogen
Manufacturer Code:PA129456
Catalog # PIPA129456
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides QLVQFDCSPELFKKIKE and KAVQVVLRICYS corresponding to amino acids 44-60 and 187-198 of Human PAIAS gamma. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 SUMO-protein ligase PIAS4; PIAS-gamma; PIASy; protein inhibitor of activated STAT PIASy; Protein inhibitor of activated STAT protein 4; Protein inhibitor of activated STAT protein gamma; protein inhibitor of activated STAT protein PIASy; protein inhibitor of activated STAT, 4; RING-type E3 ubiquitin transferase PIAS4; zinc finger, MIZ-type containing 6
Gene Aliases: PIAS-gamma; PIAS4; PIASG; PIASY; ZMIZ6
UniProt ID: (Human) Q8N2W9, (Mouse) Q9JM05
Entrez Gene ID: (Human) 51588, (Rat) 362827, (Mouse) 59004
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.